Assay Design
Gene Transcripts
- Gene Symbol
- cwINV4
- Entrez Gene ID
-
818191
- Gene Name
- cell wall invertase 4
- Gene Aliases
-
AT2G36190, AtCWIN4, AtcwINV4, F9C22.8, cell wall invertase 4
- Chromosome Location
-
Chr.2: 15174624 - 15178086 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_129177.3 |
NP_565837.1 |
5 - 6 |
1885 |
71 |
GenBank mRNA |
AB049617.1 |
- |
5 - 6 |
1584 |
71 |
View Full Product Details