Assay Design



Gene Transcripts

Gene Symbol
ABCG33
Entrez Gene ID
818305
Gene Name
pleiotropic drug resistance 5
Gene Aliases
AT2G37280, ATP-binding cassette G33, ATPDR5, F3G5.7, F3G5_7, PDR5, PLEIOTROPIC DRUG RESISTANCE 5, pleiotropic drug resistance 5
Chromosome Location
Chr.2: 15650169 - 15656657 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336645.1 NP_001325069.1 4 - 5 813 79
NM_001336647.1 NP_001325070.1 3 - 4 815 79
NM_129284.2 NP_181265.1 4 - 5 813 79

View Full Product Details