Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_129369.6 | NP_181348.1 | 4 - 5 | 555 | 98 |
NM_201900.2 | NP_973629.1 | 4 - 5 | 498 | 98 | |
GenBank mRNA | AK229007.1 | - | 4 - 5 | 450 | 98 |
BT010841.1 | - | 3 - 4 | 429 | 98 | |
BT012615.1 | - | 2 - 3 | 306 | 98 |