Assay Design



Gene Transcripts

Gene Symbol
PECT1
Entrez Gene ID
818449
Gene Name
phosphorylethanolamine cytidylyltransferase 1
Gene Aliases
AT2G38670, T6A23.13, T6A23_13, phosphorylethanolamine cytidylyltransferase 1
Chromosome Location
Chr.2: 16168712 - 16171918 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129424.5 NP_181401.1 7 - 8 1337 87
GenBank mRNA AY092984.1 - 7 - 8 1177 87
AY128807.1 - 7 - 8 1070 87

View Full Product Details