Assay Design



Gene Transcripts

Gene Symbol
ABCB6
Entrez Gene ID
818533
Gene Name
P-glycoprotein 6
Gene Aliases
AT2G39480, ATP-binding cassette B6, F12L6.14, F12L6_14, P-glycoprotein 6, PGP6
Chromosome Location
Chr.2: 16477792 - 16485039 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336781.1 NP_001324134.1 4 - 5 678 143
NM_129506.2 NP_181480.1 4 - 5 1048 143

View Full Product Details