Assay Design



Gene Transcripts

Gene Symbol
RTFL8
Entrez Gene ID
818554
Gene Name
ROTUNDIFOLIA like 8
Gene Aliases
AT2G39705, DEVIL 11, DVL11, ROTUNDIFOLIA like 8
Chromosome Location
Chr.2: 16556801 - 16558268 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129527.3 NP_565910.1 1 - 1 474 109
GenBank mRNA AK117621.1 - 1 - 1 419 109
AY086777.1 - 1 - 1 363 109

View Full Product Details