Assay Design



Gene Transcripts

Gene Symbol
ISA1
Entrez Gene ID
818580
Gene Name
isoamylase 1
Gene Aliases
ARABIDOPSIS THALIANA ISOAMYLASE 1, AT2G39930, ATISA1, T28M21.9, T28M21_9, isoamylase 1
Chromosome Location
Chr.2: 16665851 - 16672494 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336807.1 NP_001324597.1 13 - 14 1770 141
NM_001336808.1 NP_001324596.1 13 - 14 1770 141
NM_129551.4 NP_181522.1 13 - 14 1942 141
GenBank mRNA BT000443.1 - 13 - 14 1746 141
BT010348.1 - 13 - 14 1715 141

View Full Product Details