Assay Design



Gene Transcripts

Gene Symbol
CKX1
Entrez Gene ID
818749
Gene Name
cytokinin oxidase/dehydrogenase 1
Gene Aliases
AT2G41510, ATCKX1, CYTOKININ OXIDASE/DEHYDROGENASE 1, T32G6.3, T32G6_3, cytokinin oxidase/dehydrogenase 1
Chromosome Location
Chr.2: 17314412 - 17317354 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336920.1 NP_001318403.1 1 - 2 757 71
NM_001336921.1 NP_001324623.1 1 - 2 757 71
NM_129714.4 NP_181682.2 1 - 2 877 71

View Full Product Details