Assay Design



Gene Transcripts

Gene Symbol
MBF1A
Entrez Gene ID
818868
Gene Name
multiprotein bridging factor 1A
Gene Aliases
ARABIDOPSIS THALIANA MULTIPROTEIN BRIDGING FACTOR 1A, AT2G42680, ATMBF1A, F14N22.5, F14N22_5, multiprotein bridging factor 1A
Chromosome Location
Chr.2: 17774832 - 17776320 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129829.4 NP_565981.1 3 - 4 448 73
GenBank mRNA AF370280.1 - 3 - 4 372 73
AY063014.1 - 3 - 4 308 73
AY088140.1 - 3 - 4 377 73

View Full Product Details