Assay Design



Gene Transcripts

Gene Symbol
PAO2
Entrez Gene ID
818904
Gene Name
polyamine oxidase 2
Gene Aliases
AT2G43020, ATPAO2, MFL8.12, MFL8_12, polyamine oxidase 2
Chromosome Location
Chr.2: 17891503 - 17894635 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129863.3 NP_181830.1 8 - 9 1182 78
GenBank mRNA AF364952.1 - 8 - 9 1100 78
BT029025.1 - 8 - 9 740 78

View Full Product Details