Assay Design



Gene Transcripts

Gene Symbol
ARA4
Entrez Gene ID
818915
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
ARA-4, ARABIDOPSIS RAB GTPASE HOMOLOG A5C, AT2G43130, ATRAB11F, ATRABA5C, F14B2.7, RABA5C
Chromosome Location
Chr.2: 17929607 - 17931027 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129875.4 NP_181842.1 1 - 2 453 93
GenBank mRNA AK317205.1 - 1 - 2 360 93
BT025784.1 - 1 - 2 330 93
D01026.1 - - 333 93

View Full Product Details