Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202813.1 | NP_001189742.1 | 1 - 2 | 229 | 90 |
NM_129896.4 | NP_181863.1 | 1 - 2 | 237 | 90 | |
GenBank mRNA | AY065372.1 | - | 1 - 2 | 224 | 90 |
AY087030.1 | - | 1 - 2 | 222 | 90 | |
AY096479.1 | - | 1 - 2 | 156 | 90 |