Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202813.1 | NP_001189742.1 | 5 - 6 | 658 | 74 |
NM_129896.4 | NP_181863.1 | 5 - 6 | 666 | 74 | |
GenBank mRNA | AY065372.1 | - | 5 - 6 | 653 | 74 |
AY087030.1 | - | 5 - 6 | 651 | 74 | |
AY096479.1 | - | 5 - 6 | 585 | 74 |