Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001084585.2 | NP_001078054.1 | 1 - 2 | 195 | 72 |
NM_129973.3 | NP_181938.1 | 1 - 2 | 418 | 72 | |
GenBank mRNA | AF361801.1 | - | 1 - 2 | 185 | 72 |
AK226690.1 | - | 1 - 2 | 188 | 72 | |
AK227643.1 | - | 1 - 2 | 185 | 72 | |
AK316933.1 | - | 1 - 2 | 189 | 72 | |
AY050418.2 | - | 1 - 2 | 186 | 72 | |
AY113036.1 | - | 1 - 2 | 83 | 72 | |
D83531.1 | - | 1 - 2 | 186 | 72 |