Assay Design
Gene Transcripts
- Gene Symbol
- AT3G09100
- Entrez Gene ID
-
820064
- Gene Name
- mRNA capping enzyme family protein
- Gene Aliases
-
AT3G09100
- Chromosome Location
-
Chr.3: 2787953 - 2793547 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_202534.2 |
NP_974263.1 |
15 - 16 |
2092 |
126 |
View Full Product Details