Assay Design



Gene Transcripts

Gene Symbol
VSR4
Entrez Gene ID
815960
Gene Name
vacuolar sorting receptor 4
Gene Aliases
AT2G14720, BP80-2;1, MTV4, T6B13.2, VACUOLAR SORTING RECEPTOR 2, VACUOLAR SORTING RECEPTOR 2;1, VSR-2, VSR2;1, binding protein of 80 kDa 2;1, vacuolar sorting receptor 4
Chromosome Location
Chr.2: 6300488 - 6304445 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127036.5 NP_179079.1 11 - 12 2079 148
NM_179623.2 NP_849954.1 11 - 12 2018 148
GenBank mRNA AY062744.1 - 11 - 12 1957 148

View Full Product Details