Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_127036.5 | NP_179079.1 | 6 - 7 | 1606 | 178 |
NM_179623.2 | NP_849954.1 | 6 - 7 | 1545 | 178 | |
GenBank mRNA | AY062744.1 | - | 6 - 7 | 1484 | 178 |
BT008390.1 | - | 6 - 7 | 1317 | 178 |