Assay Design



Gene Transcripts

Gene Symbol
AP19
Entrez Gene ID
816245
Gene Name
associated protein 19
Gene Aliases
AT2G17380, CLATHRIN ASSEMBLY PROTEIN AP19, F5J6.14, F5J6_14, associated protein 19
Chromosome Location
Chr.2: 7552992 - 7555163 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127293.3 NP_565415.1 2 - 3 217 157
GenBank mRNA AY085130.1 - 2 - 3 207 157
BT025619.1 - 1 - 2 87 157

View Full Product Details