Assay Design
Gene Transcripts
- Gene Symbol
- CYP71A13
- Entrez Gene ID
-
817628
- Gene Name
- cytochrome P450 family 71 polypeptide
- Gene Aliases
-
AT2G30770, T11J7.16, T11J7_16, cytochrome P450, family 71, polypeptide 13, subfamily A
- Chromosome Location
-
Chr.2: 13109909 - 13111988 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128630.3 |
NP_180635.3 |
3 - 4 |
1165 |
137 |
View Full Product Details