Assay Design



Gene Transcripts

Gene Symbol
CNGC7
Entrez Gene ID
838169
Gene Name
cyclic nucleotide gated channel 7
Gene Aliases
AT1G15990, ATCNGC7, T24D18.9, T24D18_9, cyclic nucleotide gated channel 7
Chromosome Location
Chr.1: 5491133 - 5494192 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332197.1 NP_001319014.1 3 - 4 696 150

View Full Product Details