Assay Design
Gene Transcripts
- Gene Symbol
- UBQ14
- Entrez Gene ID
-
828148
- Gene Name
- Ubiquitin family protein
- Gene Aliases
-
AT4G02890, T5J8.21, T5J8_21
- Chromosome Location
-
Chr.4: 1278530 - 1280166 on Build TAIR10.1
- Gene Symbol
- UBQ10
- Entrez Gene ID
-
825880
- Gene Name
- polyubiquitin 10
- Gene Aliases
-
AT4G05320, C17L7.240, C17L7_240, UBI10, polyubiquitin 10, ubiquitin 10
- Chromosome Location
-
Chr.4: 2717977 - 2720308 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK316842.1 |
- |
2 - 2 |
972 |
120 |
AY087759.1 |
- |
2 - 2 |
972 |
120 |
- Gene Symbol
- UBQ11
- Entrez Gene ID
-
825847
- Gene Name
- ubiquitin 11
- Gene Aliases
-
AT4G05050, C17L7.6, ubiquitin 11
- Chromosome Location
-
Chr.4: 2588004 - 2589437 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BT001165.1 |
- |
2 - 2 |
676 |
120 |
View Full Product Details