Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_117252.6 | NP_192920.3 | 8 - 9 | 3654 | 117 |
NM_179208.3 | NP_849539.1 | 9 - 10 | 3558 | 117 | |
GenBank mRNA | AF138281.2 | - | 9 - 10 | 2843 | 117 |
DQ812124.1 | - | 8 - 9 | 2971 | 117 |