Assay Design
Gene Transcripts
- Gene Symbol
- LSH10
- Entrez Gene ID
-
818861
- Gene Name
- LIGHT-DEPENDENT SHORT HYPOCOTYLS-like protein (DUF640)
- Gene Aliases
-
AT2G42610, LIGHT SENSITIVE HYPOCOTYLS 10
- Chromosome Location
-
Chr.2: 17747781 - 17750524 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_180043.1 |
NP_850374.1 |
1 - 2 |
652 |
119 |
GenBank mRNA |
AK119048.1 |
- |
1 - 2 |
652 |
119 |
View Full Product Details