Assay Design



Gene Transcripts

Gene Symbol
RPP8
Entrez Gene ID
834367
Gene Name
Disease resistance protein (CC-NBS-LRR class) family
Gene Aliases
AT5G43470, DISEASE RESISTANCE PROTEIN RPP8, HRT, HYPERSENSITIVE RESPONSE TO TCV, MWF20.19, MWF20_19, RCY1, RECOGNITION OF PERONOSPORA PARASITICA 8, RESISTANT TO CMV(Y) 1
Chromosome Location
Chr.5: 17462547 - 17467448 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344485.1 NP_001318736.1 3 - 4 3188 154
NM_001344486.1 NP_001332592.1 4 - 5 3229 154
NM_180793.1 NP_851124.1 5 - 6 2914 154

View Full Product Details