Assay Design



Gene Transcripts

Gene Symbol
MYB20
Entrez Gene ID
842938
Gene Name
myb domain protein 20
Gene Aliases
AT1G66230, AtMYB20, T6J19.5, T6J19_5, myb domain protein 20
Chromosome Location
Chr.1: 24677121 - 24678580 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105294.3 NP_176797.1 1 - 2 465 141
GenBank mRNA AY519565.1 - 1 - 2 264 141
BT028904.1 - 1 - 2 264 141

View Full Product Details