Assay Design



Gene Transcripts

Gene Symbol
ARF12
Entrez Gene ID
840331
Gene Name
auxin response factor 12
Gene Aliases
AT1G34310, F23M19.4, F23M19_4, auxin response factor 12
Chromosome Location
Chr.1: 12508548 - 12511520 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103153.2 NP_174691.2 5 - 6 462 153
GenBank mRNA AB493494.1 - 5 - 6 462 153
AY669792.1 - 5 - 6 462 153

View Full Product Details