Assay Design
Gene Transcripts
- Gene Symbol
- AT3G14040
- Entrez Gene ID
-
820618
- Gene Name
- Pectin lyase-like superfamily protein
- Gene Aliases
-
AT3G14040
- Chromosome Location
-
Chr.3: 4648236 - 4650319 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112258.4 |
NP_188020.1 |
2 - 3 |
666 |
169 |
View Full Product Details