Assay Design



Gene Transcripts

Gene Symbol
BHLH32
Entrez Gene ID
822159
Gene Name
basic helix-loop-helix 32
Gene Aliases
AT3G25710, ATAIG1, TARGET OF MONOPTEROS 5, TMO5, basic helix-loop-helix 32
Chromosome Location
Chr.3: 9369499 - 9371487 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113470.3 NP_189199.1 1 - 2 591 107
GenBank mRNA AY058840.1 - 1 - 2 562 107
AY079033.1 - 1 - 2 492 107
AY086383.1 - 1 - 2 571 107
DQ205679.1 - 1 - 2 492 107

View Full Product Details