Assay Design



Gene Transcripts

Gene Symbol
AER
Entrez Gene ID
831560
Gene Name
alkenal reductase
Gene Aliases
AT-AER, AT5G16970, F2K13.120, F2K13_120, alkenal reductase
Chromosome Location
Chr.5: 5575973 - 5578086 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121703.4 NP_197199.1 4 - 5 746 114
GenBank mRNA BT022058.1 - 4 - 5 661 114
Z49768.1 - - 740 114

View Full Product Details