Assay Design
Gene Transcripts
- Gene Symbol
- HR1
- Entrez Gene ID
-
824209
- Gene Name
- homolog of RPW8 1
- Gene Aliases
-
AT3G50450, homolog of RPW8 1
- Chromosome Location
-
Chr.3: 18722234 - 18723310 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114905.2 |
NP_190614.1 |
1 - 2 |
476 |
149 |
View Full Product Details