Assay Design



Gene Transcripts

Gene Symbol
SHM4
Entrez Gene ID
827027
Gene Name
serine hydroxymethyltransferase 4
Gene Aliases
AT4G13930, DL3005C, FCAALL.160, SERINE HYDROXYMETHYLTRANSFERASE 4, serine hydroxymethyltransferase 4
Chromosome Location
Chr.4: 8047692 - 8050157 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117467.4 NP_193129.1 2 - 3 1207 159
GenBank mRNA AF361589.1 - 2 - 3 1134 159
AY093987.1 - 2 - 3 1071 159

View Full Product Details