Assay Design
Gene Transcripts
- Gene Symbol
- CYP71B14
- Entrez Gene ID
-
832589
- Gene Name
- cytochrome P450, family 71, subfamily B, polypeptide 14
- Gene Aliases
-
''cytochrome P450, AT5G25180, F21J6.102, F21J6_102, cytochrome P450, family 71, polypeptide 14, polypeptide 14'', subfamily B
- Chromosome Location
-
Chr.5: 8694630 - 8697108 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122427.2 |
NP_197900.1 |
2 - 3 |
947 |
136 |
View Full Product Details