Assay Design



Gene Transcripts

Gene Symbol
ATS9
Entrez Gene ID
839789
Gene Name
non-ATPase subunit 9
Gene Aliases
AT1G29150, REGULATORY PARTICLE NON-ATPASE 6, RPN6, non-ATPase subunit 9
Chromosome Location
Chr.1: 10180302 - 10182808 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332840.1 NP_001323322.1 2 - 2 1633 144
NM_102656.4 NP_174210.1 2 - 2 1656 144
GenBank mRNA AF370203.1 - 2 - 2 1437 144
AY230835.1 - 2 - 2 1396 144
AY230836.1 - 2 - 2 1431 144

View Full Product Details