Assay Design



Gene Transcripts

Gene Symbol
ROC2
Entrez Gene ID
824773
Gene Name
rotamase cyclophilin 2
Gene Aliases
AT3G56070, ROTAMASE CYCLOPHILIN 2, ROTAMASE CYP 2, rotamase cyclophilin 2
Chromosome Location
Chr.3: 20806566 - 20808168 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339763.1 NP_001319764.1 2 - 2 881 108
NM_115465.5 NP_191166.1 2 - 2 825 108
GenBank mRNA AY072128.1 - 2 - 2 749 108

View Full Product Details