Assay Design
Gene Transcripts
- Gene Symbol
- CYP82C3
- Entrez Gene ID
-
829325
- Gene Name
- cytochrome P450, family 82, subfamily C, polypeptide 3
- Gene Aliases
-
''cytochrome P450, AT4G31950, F11C18.9, cytochrome P450, family 82, polypeptide 3, polypeptide 3'', subfamily C
- Chromosome Location
-
Chr.4: 15455083 - 15457121 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119346.3 |
NP_194923.1 |
1 - 1 |
402 |
106 |
GenBank mRNA |
AY142533.1 |
- |
1 - 1 |
322 |
106 |
View Full Product Details