Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001341499.1 | NP_001320024.1 | 2 - 2 | 656 | 72 |
NM_118276.4 | NP_193887.1 | 3 - 3 | 760 | 72 | |
NM_202856.3 | NP_974585.1 | 2 - 2 | 1095 | 72 | |
GenBank mRNA | AY136475.1 | - | 3 - 3 | 723 | 72 |
BT006261.1 | - | 1 - 1 | 307 | 72 |