Assay Design



Gene Transcripts

Gene Symbol
FYD
Entrez Gene ID
820437
Gene Name
FYD
Gene Aliases
AT3G12570
Chromosome Location
Chr.3: 3988590 - 3991079 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035604.2 NP_001030681.1 3 - 3 1673 63
NM_112092.3 NP_566427.1 2 - 2 1586 63
NM_180240.2 NP_850571.1 2 - 2 1812 63
NM_202565.2 NP_974294.1 3 - 3 1623 63
GenBank mRNA AF360293.1 - 2 - 2 1732 63
AY070020.1 - 1 - 1 1351 63

View Full Product Details