Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035604.2 | NP_001030681.1 | 3 - 3 | 505 | 64 |
NM_112092.3 | NP_566427.1 | 2 - 2 | 418 | 64 | |
NM_180240.2 | NP_850571.1 | 2 - 2 | 644 | 64 | |
NM_202565.2 | NP_974294.1 | 3 - 3 | 455 | 64 | |
GenBank mRNA | AF360293.1 | - | 2 - 2 | 564 | 64 |
AY070020.1 | - | 1 - 1 | 183 | 64 |