Assay Design



Gene Transcripts

Gene Symbol
MLP34
Entrez Gene ID
843422
Gene Name
MLP-like protein 34
Gene Aliases
AT1G70850, F15H11.10, F15H11_10, MLP-like protein 34
Chromosome Location
Chr.1: 26714866 - 26716901 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036188.1 NP_001031265.1 3 - 3 1054 144
NM_105753.4 NP_565003.3 3 - 3 1176 144
NM_180645.3 NP_850976.1 3 - 3 1100 144
GenBank mRNA AF372899.1 - 3 - 3 1054 144
AK230234.1 - - 3367 144

View Full Product Details