Assay Design



Gene Transcripts

Gene Symbol
PAC1
Entrez Gene ID
821774
Gene Name
20S proteasome alpha subunit C1
Gene Aliases
20S PROTEASOME SUBUNIT PAC1, 20S proteasome alpha subunit C1, AT3G22110
Chromosome Location
Chr.3: 7792549 - 7794262 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113108.5 NP_188850.1 2 - 2 943 76
GenBank mRNA AF043521.1 - 2 - 2 810 76
AF386953.1 - 2 - 2 835 76
AY085914.1 - 2 - 2 792 76

View Full Product Details