Assay Design
Gene Transcripts
- Gene Symbol
- RALFL11
- Entrez Gene ID
-
816420
- Gene Name
- ralf-like 11
- Gene Aliases
-
AT2G19030, T20K24.4, T20K24_4, ralf-like 11
- Chromosome Location
-
Chr.2: 8247726 - 8248181 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127460.2 |
NP_179493.1 |
1 - 1 |
208 |
158 |
View Full Product Details