Assay Design
Gene Transcripts
- Gene Symbol
- ACT3
- Entrez Gene ID
-
824542
- Gene Name
- actin 3
- Gene Aliases
-
AT3G53750, actin 3
- Chromosome Location
-
Chr.3: 19915257 - 19917775 on Build TAIR10.1
- Gene Symbol
- ACT1
- Entrez Gene ID
-
818339
- Gene Name
- actin 1
- Gene Aliases
-
AAc1, ARABIDOPSIS ACTIN 1, AT2G37620, F13M22.12, F13M22_12, actin 1
- Chromosome Location
-
Chr.2: 15778941 - 15781561 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY049293.1 |
- |
4 - 4 |
1290 |
118 |
View Full Product Details