Assay Design



Gene Transcripts

Gene Symbol
SCRL4
Entrez Gene ID
3767592
Gene Name
SCR-like 4
Gene Aliases
AT1G60986, SCR-like 4
Chromosome Location
Chr.1: 22459000 - 22459650 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036136.2 NP_001031213.1 1 - 2 148 65
GenBank mRNA DQ912181.1 - 1 - 2 76 65

View Full Product Details