Assay Design



Gene Transcripts

Gene Symbol
LCR1
Entrez Gene ID
3771464
Gene Name
low-molecular-weight cysteine-rich 1
Gene Aliases
AT5G48543, low-molecular-weight cysteine-rich 1
Chromosome Location
Chr.5: 19674358 - 19675465 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036957.2 NP_001032034.1 1 - 2 94 139

View Full Product Details