Assay Design



Gene Transcripts

Gene Symbol
KCMF1
Entrez Gene ID
613522
Gene Name
potassium channel modulatory factor 1
Gene Aliases
-
Chromosome Location
Chr.11: 49939796 - 50018771 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076275.1 NP_001069743.1 3 - 4 410 96
GenBank mRNA BC116058.1 - 3 - 4 410 96

View Full Product Details