Assay Design



Gene Transcripts

Gene Symbol
KIN
Entrez Gene ID
519670
Gene Name
Kin17 DNA and RNA binding protein
Gene Aliases
-
Chromosome Location
Chr.13: 16109948 - 16118705 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101099.1 NP_001094569.1 2 - 3 246 86
GenBank mRNA BC146082.1 - 2 - 3 246 86

View Full Product Details