Assay Design



Gene Transcripts

Gene Symbol
ERO1B
Entrez Gene ID
514361
Gene Name
endoplasmic reticulum oxidoreductase 1 beta
Gene Aliases
ERO1LB
Chromosome Location
Chr.28: 8905557 - 8977596 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206730.1 NP_001193659.1 6 - 7 500 88

View Full Product Details