Assay Design



Gene Transcripts

Gene Symbol
PPIF
Entrez Gene ID
414346
Gene Name
peptidylprolyl isomerase F
Gene Aliases
CyP-D, CypD
Chromosome Location
Chr.28: 34966787 - 34972884 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001597.1 NP_001001597.1 4 - 5 420 63
GenBank mRNA AY594335.1 - 4 - 5 420 63

View Full Product Details