Assay Design



Gene Transcripts

Gene Symbol
MTHFR
Entrez Gene ID
497032
Gene Name
methylenetetrahydrofolate reductase
Gene Aliases
-
Chromosome Location
Chr.16: 41872982 - 41888935 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011685.1 NP_001011685.1 2 - 3 178 100
GenBank mRNA AY854631.1 - 2 - 3 178 100

View Full Product Details