Assay Design



Gene Transcripts

Gene Symbol
HFE
Entrez Gene ID
497622
Gene Name
homeostatic iron regulator
Gene Aliases
-
Chromosome Location
Chr.23: 31855157 - 31864399 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012399.1 NP_001012399.1 4 - 5 979 55
GenBank mRNA AY898807.1 - 4 - 5 979 55

View Full Product Details